Order Entry
Germany
ContactUsLinkComponent
6-Carboxy-X-rhodamine succinimidyl ester (6-ROX SE) amine-reactive fluorescent dye
6-Carboxy-X-rhodamine succinimidyl ester (6-ROX SE) amine-reactive fluorescent dye
Catalog # 90037.
Supplier:  Biotium
CAS Number:  
undefined
6-Carboxy-X-rhodamine succinimidyl ester (6-ROX SE) amine-reactive fluorescent dye
Catalog # 90037.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :631.685 g/mol
Storage Temperature:Freezer
CAS Number:216699-36-4
Formula:C₃₇H₃₃N₃O₇