Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Pharmaceutical secondary standard, Ammonium chloride, (CRM), Supelco®
Pharmaceutical secondary standard, Ammonium chloride, (CRM), Supelco®
Pharmaceutical secondary standard, Ammonium chloride, (CRM), Supelco®
Catalog # PHR1327-1G
Supplier:
Merck
CAS Number:
Certificates
Expand image
Pharmaceutical secondary standard, Ammonium chloride, (CRM), Supelco®
Catalog # PHR1327-1G
Supplier:
Merck
CAS Number:
Certificates
Specifications
Pk:
1 g
CAS No.:
12125-02-9
Description:
Ammonium chloride
Description:
Ammonium chloride
Specifications
About this item
Product Details
Documentation
Product Details
Documentation