Supelco® Drug and metabolite standards, Risperidone-D4 solution, (CRM)
R-013-1ML
: Merck
:
- Concentration:100 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:1020719-76-9
- Description:Risperidone-D4 solution
- Description:Risperidone-D4 solution
- Pk Info:ampoule of 1 ml