Order Entry
Germany
ContactUsLinkComponent
Supelco® Drug and metabolite standards, Posaconazole-D4 solution, (CRM)
Supelco® Drug and metabolite standards, Posaconazole-D4 solution, (CRM)
Catalog # P-108-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Drug and metabolite standards, Posaconazole-D4 solution, (CRM)
Catalog # P-108-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1,0 mg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    1133712-26-1
  • Description:
    Posaconazole-D4 solution
  • Description:
    Posaconazole-D4 solution

Specifications

About this item