Supelco® Drug and metabolite standards, N-desmethylzopiclone-D4 solution, (CRM)
Catalog # D-112-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- Description:N-Desmethylzopiclone-D4 solution