Supelco® Drug and metabolite standards, 9-Hydroxyrisperidone-D4 solution, (CRM)
Catalog # H-095-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:1020719-55-4
- Description:9-Hydroxyrisperidone-D4 solution
- Description:9-Hydroxyrisperidone-D4 solution
- Pk Info:ampoule of 1 ml