Drug and metabolite standards, (+/-)-threo-ritalinic acid-D4 hydrochloride solution, (CRM), Supelco®
Catalog # R-025-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- Description:(+/-)-threo-Ritalinic acid-D4 hydrochloride solution