Heating mantle, KM-MPE series
: MOHR
Metal-cased heating mantle with built-in controller for operating temperatures up to 450 °C.
- Flexible glass yarn heating element provides even heat transfer and gentle treatment of the glassware
- Chemical resistant metal housing
- Thermally insulated and earthed
- Predrilled threaded hole for clamp KM-SK
- Built-in heating zone switch
System of protection: Casing IP 43, heating surface IP 00.