Multi-element standard, Varian tuning solution
Multi-element standard, 15 elements, 5% HNO₃, Safety plastic bottle, DIN45, 500 mL
Catalog # KRAF27704.2000
Supplier: Bernd Kraft
Multi-element standard, Varian tuning solution
Catalog # KRAF27704.2000
Supplier: Bernd Kraft
CAS Number:
Jump to:
Product Details & Documents
This multi-element standard of 1 litre contains 500 mg: K and 50 mg: Al, As, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn.
Specifications
Specifications
- Matrix:5% HNO₃
- Pack type:Safety plastic bottle, DIN45
- Pk:500 mL
- Description:Multi-element standard, 15 elements



