Order Entry
Germany
Orders LinkContactUsLinkComponent
Glycine buffer, Sigma-Aldrich®

Glycine buffer, Sigma-Aldrich®

Glycine buffer, 9,2 (25 °C), 100 mL
Catalog # SIALG5418-100ML
Supplier:   Merck
Glycine buffer, Sigma-Aldrich®
Catalog # SIALG5418-100ML
Supplier:  Merck
CAS Number:  
Glycine buffer, Sigma-Aldrich®

Glycine buffer, 9,2 (25 °C), 100 mL

CAS Number:  
Price
91,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

For use in the enzymatic determination of lactate using lactic dehydrogenase.

pH: 9,10 - 9,30
Storage temperature: 2 to 8 °C

Contains: Glycine 0,6 M, hydrazine 0,5 M and chloroform 0,0375 %.

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Glycine buffer
  • pH value:
    9,2 (25 °C)
  • Cat. No.:
    SIALG5418-100ML
  • M-Clarity quality level:
    MQ200

Specifications