pET-14b DNA, Novagen®
Catalog # 69660-3
Supplier: Merck Millipore (Novagen)
CAS Number:
Specifications
- Pk:10 µG
- Description:pET-14b DNA, Novagen®
Specifications
About this item
The pET-14b vector carries an N-terminal His·Tag® sequence followed by a thrombin cleavage site and three cloning sites.
- Packed in a plastic ampoule
- Store at ≤–70 °C
Delivery: The pET vectors are supplied as purified plasmid DNA (10 µg). It includes an induction control strain (supplied as a glycerol stock).