Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride ≥97% (by HPLC), white powder, Sigma-Aldrich®
: Merck
:2645-08-1
:
:C₁₅H₂₂N₄O₃
:MFCD00012579
:342.82 g/mol
:M-Clarity™ quality level = MQ200
:220-157-0
- Appearance (Color):White
- Appearance (Form):Powder
- Purity (HPLC Area %):≥97%
- Solubility (Color):Colorless
- Solubility (Turbidity):Clear
- Solubility (Method):50 mg/ml in Water
- Water:≤1%
- Suitability Assay:Suitable as Substrate for Trypsin