Industrial gloves, uvex profi ergo, ENB20A
Supplier: UVEX
Certificates
About this item
These cotton interlock safety gloves feature a special NBR coating on palm, fingers and ³/₄ of the back of the hand. Ideal for use in oily and greasy areas of application.
- Very good grip in damp, wet and oily areas
- Good tactile feel
- Ergonomic fit, highly flexible
- Also suitable for operating touch displays
- Lining provides high wearer comfort, feels pleasant against the skin, and provides optimum regulation of the glove climate
Typical applications include: Assembly, inspection, repair work, shipping/logistics and packaging.
Certifications: EN 388:2016 (2121X)
OEKO-TEX® Standard 100
OEKO-TEX® Standard 100
Specifications
- Catalog No:
- UVEX6014700
- UVEX6014701
- UVEX6014704
- UVEX6014702
- UVEX6014703
- Model:
- ENB20A
- ENB20A
- ENB20A
- ENB20A
- ENB20A
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- Food contact approved:
- +
- +
- +
- +
- +