Gel filtration media, Sephadex® G-50
Supplier: Merck
Certificates
About this item
Sephadex® G-50 is a gel filtration medium used in affinity chromatography, protein chromatrography and gel filtration chromatography.
- Swelling from 1 g to (9 to 11 ml)
It has been used to investigate effects from snake venom, isolate lectins from various species and to demonstrate the positive influence of 20% ethanol on proteinase desorption.
Packaging: Packed in poly bottle.