L(+)-Ascorbic acid 99+%, powder for electrophoresis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:50-81-7
:
Formula:C₆H₈O₆
MDL Number:MFCD00064328
:176.13 g/mol
Merck Index:14,830
Melting Pt:190…193 °C (dec.)
Density:1.65
Storage Temperature:Ambient
EINECS:200-066-2
Specification Test Results
- Assay:Min. 99%
- Form:Powder