ContactUsLinkComponent
Cut resistant gloves, Showa 4561
Cut resistant gloves, Showa 4561
Supplier:  Showa
Certificates
undefined
Cut resistant gloves, Showa 4561
Supplier:  Showa
Specifications
  • Catalog No:
  • 112-5839
  • 112-5841
  • 112-5840
  • 112-5838
  • 112-5842
  • Model:
  • 4561
  • 4561
  • 4561
  • 4561
  • 4561
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Liner material:
  • Spandex/Kevlar®
  • Spandex/Kevlar®
  • Spandex/Kevlar®
  • Spandex/Kevlar®
  • Spandex/Kevlar®
  • Finish:
  • Rough
  • Rough
  • Rough
  • Rough
  • Rough
  • Gauge:
  • 15
  • 15
  • 15
  • 15
  • 15
  • Cuff style:
  • Knitted elasticated wrist and cuff
  • Knitted elasticated wrist and cuff
  • Knitted elasticated wrist and cuff
  • Knitted elasticated wrist and cuff
  • Knitted elasticated wrist and cuff
  • EN norm:
  • EN 388:2016 (4X32D)
  • EN 388:2016 (4X32D)
  • EN 388:2016 (4X32D)
  • EN 388:2016 (4X32D)
  • EN 388:2016 (4X32D)