General purpose gloves, Showa 310
Supplier: Showa
Certificates
About this item
These hand-specific cotton gloves with latex coating on palm and fingertips are flexible and robust offering great dexterity, outstanding grip and good resistance to tearing. They are ideal for heavy duty manual handling.
- Latex coating provides protection against abrasions, alcohols and aggressive detergents
- Excellent grip in damp and dry environments
- Seamless knit to prevent irritation
- Breathable back of hand to reduce perspiration
- Designed for easy movement and continuous wear thanks to renowned ergonomic shaped hand mould
Ideal for use in construction, highway maintenance, logistics, municipal services, heavy engineering. Typical applications include: Bricklaying, maintenance, manual handling, refuse collection and tiling.
Certifications: EN 388:2016 (2142X); Cat. II.
Specifications
- Catalog No:
- 112-5773
- 112-5774
- 112-5778
- 112-5777
- 112-5775
- 112-5779
- 112-5767
- 112-5768
- 112-5769
- 112-5770
- 112-5776
- 112-5772
- 112-5771
- Model:
- 310G
- 310G
- 310OR
- 310OR
- 310G
- 310OR
- 310B
- 310B
- 310B
- 310B
- 310OR
- 310G
- 310G
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Liner material:
- Cotton/polyester
- Cotton/polyester
- Cotton/polyester
- Cotton/polyester
- Cotton/polyester
- Cotton/polyester
- Cotton/polyester
- Cotton/polyester
- Cotton/polyester
- Cotton/polyester
- Cotton/polyester
- Cotton/polyester
- Cotton/polyester
- Finish:
- Palm-dipped natural rubber coating
- Palm-dipped natural rubber coating
- Palm-dipped natural rubber coating
- Palm-dipped natural rubber coating
- Palm-dipped natural rubber coating
- Palm-dipped natural rubber coating
- Palm-dipped natural rubber coating
- Palm-dipped natural rubber coating
- Palm-dipped natural rubber coating
- Palm-dipped natural rubber coating
- Palm-dipped natural rubber coating
- Palm-dipped natural rubber coating
- Palm-dipped natural rubber coating
- Gauge:
- 10
- 10
- 10
- 10
- 10
- 10
- 10
- 10
- 10
- 10
- 10
- 10
- 10
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (2142X)
- EN 388:2016 (2142X)
- EN 388:2016 (2142X)
- EN 388:2016 (2142X)
- EN 388:2016 (2142X)
- EN 388:2016 (2142X)
- EN 388:2016 (2142X)
- EN 388:2016 (2142X)
- EN 388:2016 (2142X)
- EN 388:2016 (2142X)
- EN 388:2016 (2142X)
- EN 388:2016 (2142X)
- EN 388:2016 (2142X)