ContactUsLinkComponent
General purpose gloves, Showa 350R
General purpose gloves, Showa 350R
Supplier:  Showa
Certificates
undefined
General purpose gloves, Showa 350R
Supplier:  Showa
Specifications
  • Catalog No:
  • 112-5788
  • 112-5789
  • 112-5790
  • 112-5791
  • Model:
  • 350R
  • 350R
  • 350R
  • 350R
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Liner material:
  • Polyester/cotton
  • Polyester/cotton
  • Polyester/cotton
  • Polyester/cotton
  • Finish:
  • Nitrile palm coating
  • Nitrile palm coating
  • Nitrile palm coating
  • Nitrile palm coating
  • Gauge:
  • 10
  • 10
  • 10
  • 10
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4132X)
  • EN 388:2016 (4132X)
  • EN 388:2016 (4132X)
  • EN 388:2016 (4132X)
Specifications