Protein A/G magnetic agarose beads, Pierce™
Supplier: Thermo Fisher Scientific
Certificates
About this item
Protein A/G magnetic agarose beads provide a fast, convenient method for purification of immunoglobulins from serum, cell culture supernatant, or ascites.
- Sufficient for both routine and demanding purification procedures
- Optimised for use with high-throughput magnetic platforms
- Flexible for IgG binding domains of both Protein A and Protein G on one bead
- Beads are compatible with manual and automated applications
These beads are a mixture of Protein A magnetic beads and Protein G magnetic beads. The recombinant chimeric Protein A/G combines four IgG binding domains from Protein A and two binding domains from Protein G, making it a more general and convenient tool for purifying immunoglobulins.
Delivery: Supplied as 20 ml of 25% v/v suspension in phosphate buffered saline, 0,01% Tween-20 detergent, 0,05% sodium azide.