Croscarmellose sodium NF
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:74811-65-7
:
Formula:C₂₈H₃₀Na₈O₂₇
MDL Number:MFCD00217760
:982.44 g/mol
Storage Temperature:Ambient
Specification Test Results
- pH of a 1% Dispersion:5 - 7
- Degree of Substitution (Dried Basis):0.6 - 0.85
- Residue on Ignition:14 - 28.00%
- Settling Volume:To Pass Test
- Loss on Drying:10.0%
- Sodium Chloride and Sodium Glycolate:0.5%
- Content of Water-Soluble Material:10%
- Microbial Limits:To Pass Test
- Cadmium (Cd):As reported
- Lead (Pb):As reported
- Arsenic (As):As reported
- Mercury (Hg):As reported
- Copper (Cu):As reported
- Identification:To Pass Test
- Residual Solvents:To Pass Test