Order Entry
Germany
ContactUsLinkComponent
 
Boc-Trp(Boc)-OH ≥98% (by TLC, NMR), Novabiochem®
  04-12-0205-25
 :  
Boc-Trp(Boc)-OH ≥98% (by TLC, NMR), Novabiochem®
  04-12-0205-25
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Colour (Visual):
    White to Slight Yellow to Beige
  • Appearance of Substance (Visual):
    Powder
  • Identity (IR):
    Passes Test
  • Optical Rotation Α 25/D (C=1 in Methanol):
    +13.5 - +17.5°
  • Purity (TLC(011A)):
    ≥98%
  • Purity (TLC(0811)):
    ≥98%
  • Solubility (1 mmole in 2 ml DMF):
    Clear Solution

 

 

 :
 :404.463 g/mol
 :Ambient
 :144599-95-1
 :C₂₁H₂₈N₂O₆
 :MFCD00270520