

Crystal violet solution R Reag. Ph. Eur. chapter 4.1.1
Catalog # KRAF27773.0700
Supplier: Bernd Kraft
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:407.99 g/mol
UN:2789
ADR:3.8,II
:GHS05
:GHS02
CAS Number:548-62-9
Formula:C₂₅H₃₀ClN₃
MDL Number:MFCD00011750