Sodium hydroxide solution 35% for analysis
Plastic canister, UN-approved, 5 L
Catalog # KRAF20047.5020
Supplier: Bernd Kraft
Sodium hydroxide solution 35% for analysis
Catalog # KRAF20047.5020
Supplier: Bernd Kraft
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- UN:1824
- ADR:8,II
Specifications
Specifications
- Pack type:Plastic canister, UN-approved
- Pk:5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



