Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fosmidomycin sodium salt
Fosmidomycin sodium salt
Fosmidomycin sodium salt
Fosmidomycin sodium salt
Catalog # MOLE11512216-10MG
Supplier:
Molekula
CAS Number:
Expand image
Fosmidomycin sodium salt
Catalog # MOLE11512216-10MG
Supplier:
Molekula
CAS Number:
Specifications
Pk:
10 mg
Seq:
2275
Specifications
About this item
Product Details
:
:
205.1 g/mol
CAS Number:
66508-37-0
Formula:
C₄H₉NO₅P·Na
Product Details