Order Entry
Germany
ContactUsLinkComponent
 
Fmoc-Leu-Ser(psiMe,Mepro)-OH, Novabiochem®
 
Fmoc-Leu-Ser(psiMe,Mepro)-OH, Novabiochem®

 

  • CAS No.:
    339531-50-9
  • Protein synonyms:
    Fmoc-Leu-Ser(psiMe,Mepro)-OH|(S)-3-(N-Fmoc-L-leucinyl)-2,2-dimethyloxazolidine-4-carboxylic acid
  • Protein/peptide name:
    Fmoc-Leu-Ser(psiMe,Mepro)-OH
  • Purity:
    97.0%
  • Molecular weight:
    480.55