Vacuum gauge
Catalog # NEUB2283-10-1000
Supplier: NEUBERT VOLUME GLASSWAERE
Specifications
- Measurement range:0 - 1000 mbar
- Description:Vacuum gauge
- Pk:1
Specifications
About this item
It is complete with adapter on core NS 29/32 and suitable for work in the rough vacuum range.