Graduated pipettes for enzyme test, Class AS
Graduated pipettes with long pulled out tip, ring divisions and partial sequence above O-point.
: DIN 12699; Class AS, conformity certified.
Graduated pipettes with long pulled out tip, ring divisions and partial sequence above O-point.