LC columns, AdvanceBio RP-mAb, C4
: Agilent
AdvanceBio RP-mAb columns are designed to optimise the performance of intact and reduced mAb analysis when analysing monoclonal antibodies for biopharma discovery, development, and QA/QC applications.
- Superficially porous particles (3,5 µm) with wide pores (450 Å) increase mAb resolution while maintaining compatibility with all LC instruments
- Shorter analysis times compared to columns packed with fully porous particles of the same size
- Robust poroshell packed bed and 2 µm inlet frit extend column lifetime by helping prevent inlet blockage
The wide-pore poroshell technology of the column delivers high efficiency, a short analysis time, and low pressure, at temperatures below 80 °C.
- Catalog No:
- HEWL793775-904
- HEWL799975-904
- HEWL799775-904
- HEWL797775-904
- HEWL795975-904
- HEWL793975-904
- HEWL795775-904
- Max. pressure:
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- pH range:
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- Temperature limit:
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- USP code:
- L26
- L26
- L26
- L26
- L26
- L26
- L26