Safety goggles, 6X1, perfect hybrid
Catalog # UNVT6X1.00.00.00
Supplier: UNIVET
Specifications
- Type:Anti-UV, indirectly vented, anti-fog, anti-scratch
- Colour:Green
- Seq:02
- Lenses:Clear PC
- Pk:5
Specifications
About this item
6X1 combines the lightness of a spectacle to the benefits of a mask. The adjustable temples with SoftPad technology can be easily replaced with the elastic band included.
- Gun metal frame
- Wide lens for a field of view without distortion
- Temples adjustable in length and inclination with SoftPad technology included for absolute versatility
- Sophisticated indirect ventilation system
- Soft internal rubber for comfort without local pressure
Provides protection against drops, sprays and large dust particles guaranteed by the special design of the internal rubber.
Weight: 54 g
Certifications: EN 166-3; EN 166-4
Delivery: Available with RX insert for corrective lenses.