Autoclavable cleanroom goggles, 619
Supplier: UNIVET
Certificates
About this item
These cleanroom googles are available with a direct or an indirect ventilation system. Ideal for use in rooms with contamination control (clean and white room) as well as in aseptic environments.
- Polycarbonate lenses protect against impacts and UV radiations
- Spherical, panoramic lens: Goggles can be worn over prescription glasses
- Elastic strap in antistatic silicone has an innovative quick adjustment system
- Indirect ventilation system in the lower part prevents the escape of particulate matter
- Can be used for prolonged periods as it does not create distortion or fatigue
- Sterilisable with steam, gamma or beta radiations
Certifications: EN 166