Tris(hydroxypropyl)phosphine solution 0.5 M, Millipore®
Supplier: Merck Millipore (Novagen)
Certificates
About this item
CAS Number:4706-17-6
:
Formula:C₉H₂₁O₃P
MDL Number:MFCD00269835
:208.24 g/mol
Storage Temperature:Freezer
UN:2922
ADR:8 (6.1),II
Specification Test Results
- Purity:>80%