Medical thermometer, Thermoval® standard
Catalog # HART9250215
Supplier: HARTMANN PAUL
Specifications
- Description:Medical thermometer, Thermoval® standard
- Pk:1
Specifications
About this item
Digital thermometer for oral, axillary and rectal measurements of the body temperature.
- Measuring time: approx. 60 - 90 seconds
- Easy to read digital display
- Automatic power-off
- Displays the last measured value
- With replaceable long-term battery
- Acoustic signal indicates when thermometer is ready for operation and when the measurement is completed
Delivery: Supplied with protective cover.