Order Entry
Germany
Orders LinkContactUsLinkComponent
Pyrase strips

Pyrase strips

Pyrase strips (50 strips + 1,0 ml reagent), 50
Catalog # SIAL67886-1KT
Supplier:   Merck
Pyrase strips
Catalog # SIAL67886-1KT
Supplier:  Merck
CAS Number:  
Pyrase strips

Pyrase strips (50 strips + 1,0 ml reagent), 50

CAS Number:  
Price
66,70 €
Log in for availability
1 * 50 ST
Product Details & Documents

This kit contains plastic strips with a paper zone saturated with chromogenic substrate for the detection of pyrrolidonyl peptidase and a bottle containing a developing reagent.

  • Result of the reaction is read after 2 min.
  • Detection of pyrase is a sensitive, reliable and rapid method of wide application

Pyrase Strips are used for diagnostic test for the rapid differentiation of Enterococci from the group D Streptococci and differentiation of Streptococcus pyogenes from other haemolytic Streptococci. The Strip can also be used for the identification of Gram-positive genera, catalase negative cocci, and coccobacilli.

Llimited shelf life, expiry date on the label.

Storage temperature: 2 - 8 °C

Delivery information: The kit contains 50 test strips and 1,0 ml reagent.

Specifications

Specifications

  • Pk:
    50
  • Description:
    Pyrase strips (50 strips + 1,0 ml reagent)
  • Cat. No.:
    SIAL67886-1KT
  • M-Clarity quality level:
    MQ100

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.