Order Entry
Germany
ContactUsLinkComponent
 
Autoclavable Bags, Biohazard
Catalog # RATI7003220
Supplier:  RATIOLAB
undefined
Autoclavable Bags, Biohazard
Catalog # RATI7003220
Supplier:  RATIOLAB

Specifications

  • Capacity:
    32 L
  • Colour:
    Red
  • L×W:
    600×800 mm
  • Material:
    PP
  • Thickness:
    50 µm
  • Pk:
    500

Specifications

About this item

Autoclavable PP bags for the disposal of contaminated labware.

  • Distinctive red colour for easy identification
  • Rectangle white indicator patch field turns into dark when autoclaved
  • BIOHAZARD symbol and safety imprint in German, English and French language
  • Autoclavable at 134 °C (20 minutes)
Caution: The bag must not be completely closed when autoclaving.