Poly(ethylene glycol) methyl ether-block-poly(lactide-co-glycolide) PEG ∽Mn 5,000, PLGA ∽Mn 7,000, Sigma-Aldrich®
Catalog # SIAL765139-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Refrigerator
Formula:H[(C₃H₄O₂)x(C₂H₂O₂)y]mO[C₂H₄O]nCH₃