ContactUsLinkComponent
Test strips for semi-quantitative analysis of low water hardness, AQUADUR® sensitive
Test strips for semi-quantitative analysis of low water hardness, AQUADUR® sensitive
Catalog # MANA91243
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test strips for semi-quantitative analysis of low water hardness, AQUADUR® sensitive
Catalog # MANA91243
Supplier:  MACHEREY-NAGEL
CAS Number:  
Specifications
  • Pk:
    50
  • Description:
    Aquadur® sensitive test strips, box, box 6×95 mm
  • Range:
    < 0,5 - >18,8 mg/l CaCO₃
Specifications
About this item
AQUADUR® sensitive test strips for semi-quantitative analysis of low water hardness, after amongst other things the softening of water.
Delivery: Supplied in one aluminium container with 50 test strips.