GC septa, thermolite® PLUS
Supplier: Restek
Certificates
About this item
These septa are preconditioned, GC-FID tested and ready to use.
- Usable to 350 °C inlet temperature
- Ultra-low bleed minimises background signal
- Plasma coating eliminates sticking
- Precision molded for consistent, accurate fit
Frequently Bought Together