Nα-(p-Toluenesulphonyl)-DL-lysine chloromethyl ketone hydrochloride ≥95% (by elemental analysis), off-white solid, Calbiochem®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
CAS Number:4238-41-9
:
Formula:C₁₄H₂₂Cl₂N₂O₃S
MDL Number:MFCD00012513
:369.31 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity (Elemental Analysis):≥95%