Seal test, Stericlin®
Supplier: VP Medical Packaging
Certificates
About this item
The Stericlin® Seal Test is used for routine testing of seal seams according to DIN EN ISO 11607. In combination with the seal seam integrity test it offers the most comprehensive and reliable routine examination for the heat sealing process.
- Clear result
- Show irregularities and damage
For specifications, see the DGSV guidelines for the validation of packaging processes according to DIN EN ISO 11607-2.