Ethanol denatured (denatured with 1% MEK, 1% IPA and 10 ppm Bitrex®)
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:64-17-5
:
Formula:H₃CCH₂OH
MDL Number:MFCD00003568
:46.07 g/mol
Boiling Pt:78.3 °C
Melting Pt:–114.5 °C
Density:0.789 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Assay (GC, Denaturant not Included):Min. 99.8%
- Acidity (as CH₃COOH):Max. 1.0 mg/100 ml
- Identity (IR):Passes Test
- Water (K.F.):Max. 0.3%