Order Entry
Germany
ContactUsLinkComponent
Tritirachium album proteinase K
Tritirachium album proteinase K
Supplier:  Merck
Certificates
Tritirachium album proteinase K
Supplier:  Merck
Certificates

Specifications

  • Species:
    Tritirachium album
  • Storage conditions:
    2...8 °C
  • Enzyme name:
    Proteinase K
  • Enzyme activity:
    ≥600 mAnson u/ml
  • pH:
    7,5

Specifications

Related Information