

Sodium selenite 98.5-101.5% (by anhydrous basis) Ph. Eur., BP, pharma grade
Catalog # APPC192756.1211
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pk:1.000 g
- Seq:3047
Specifications
Specification Test Results
- Assay (Calculated on Anhydrous Basis):98.5 - 101.5%
- Identity According to Pharmacopoeias:Passes Test
- pH of 10% Solution:9.8 - 10.8
- Loss on Drying at 130 °C:≤0.5%
- Chloride (Cl):≤0.005%
- Sulphate and Selenates (as SO₄):≤0.03%
- Fe:≤0.005%
Specification Test Results
About this item
:
ADR:6.1,I
:172.94 g/mol
EINECS:233-267-9
UN:2630
:GHS06
:GHS09
Storage Temperature:Ambient
CAS Number:10102-18-8
Formula:Na₂SeO₃
MDL Number:MFCD00003489