Seal seam integrity test, Stericlin®
Supplier: VP Medical Packaging
Certificates
About this item
Stericlin® seal seam integrity tests are used for routine testing of seal seams according to DIN EN ISO 11607. In combination with the Seal Test they offer comprehensive and secure routine testing for the heat sealing process.
- Easy to perform
- Quickly detects channels and voids
For specifications, see the DGSV guidelines for the validation of packaging processes according to DIN EN ISO 11607-2.
Ordering information: Seal seam integrity tests are available in two basic versions: 'Pipette' and 'disposable ink bag'.