Human IgA purification kit, LigaTrap®
Supplier: LIGATRAP TECHNOLOGIES
Certificates
About this item
LigaTrap® Human IgA Purification Kit with LT technology utilises novel, patented ligands, designed for IgA antibody purification. Kit provides all the necessary reagents for fast, convenient micro-scale purification of human IgA in just ten easy steps.
- Designed to purify human IgA
- For in vitro laboratory use
- Binding capacity for monoclonal human IgA is >15 mg/ml resin
- Storage: 2 to 8 °C
LigaTrap® Human IgA purification resin is engineered to purify high quality human IgA antibodies from recombinant and monoclonal sources. Resins bind an exceptionally broad range of recombinant, monoclonal and human/animal source polyclonal immunoglobulins.
Each spin column can be used, regenerated, and used up to ten times with minimal
loss in binding capacity.
Delivery: Each kit contains ten microspin columns prefilled with 0,10 ml of LigaTrap® human IgA purification resin, buffers and collection tubes.
Caution: Not for diagnostic or therapeutic use. Not for human or animal consumption.