ContactUsLinkComponent
Sample loops for HPLC injector, Rheodyne 7725/7725i
Sample loops for HPLC injector, Rheodyne 7725/7725i
Catalog # SIAL57628-U
Supplier:  Merck
Sample loops for HPLC injector, Rheodyne 7725/7725i
Catalog # SIAL57628-U
Supplier:  Merck
Specifications
  • Capacity:
    200 µl
  • Description:
    Sample loop with fittings
  • Seq:
    08
  • Pk:
    1