ContactUsLinkComponent
Glycan BEH amide columns, ACQUITY UPLC™
Glycan BEH amide columns, ACQUITY UPLC™
Supplier:  WATERS
Certificates
Glycan BEH amide columns, ACQUITY UPLC™
Supplier:  WATERS
Certificates
Specifications
  • Catalog No:
  • WATE186004740
  • WATE186004741
  • WATE186004742
  • Carbon load:
  • 12%
  • 12%
  • 12%
  • pH range:
  • 2 - 11
  • 2 - 11
  • 2 - 11
  • Surface area:
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g