Nitrile gloves, SemperCare®
The ambidextrous disposable SemperCare® Nitrile combines superb barrier protection, comfort, fit, and feel while providing the tensile strength necessary for demanding environments.
- Easy donning, improved fit and comfort
- Strong and more durable
- Powder- and latex-free
- Beaded cuff
- Textured fingers and palm for safe grip, even in wet environments
- Smart Dispense Technology, Single Glove Delivery® for fewer wrinkles
Appropriate for handling cytotoxic chemicals. Safe for use with sensitive electronics.
- Catalog No:
- SEMP826781639
- SEMP826781637
- SEMP826781635
- SEMP826781633
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured fingers and palm
- Textured fingers and palm
- Textured fingers and palm
- Textured fingers and palm