Order Entry
Germany
ContactUsLinkComponent
GC-FID test mix (3 components)
GC-FID test mix (3 components)
Catalog # REST35108
Supplier:  Restek
CAS Number:  
undefined
GC-FID test mix (3 components)
Catalog # REST35108
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    20 µg/mleach in hexane
  • Pk:
    1
  • Description:
    GC-FID test mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The mix consists of (C12) n-dodecane, (C14) n-tetradecane and (C16) n-hexadecane.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.