BAC resolution control standard t-B (6 components)
Artikelnr. REST36011
Supplier: Restek
CAS Number:
Specifications
- Concentration:100 mg/dl each in water
- Pk:1 Ampoul
- Description:BAC resolution control standard t-B
- Pk Info:1 ml/ampoule
Specifications
About this item
The mix is used to verify the retention time for each compound normally included in a blood alcohol test, and to verify that the compounds are resolved from and do not interfere with one another.
- Includes tert-butanol internal standard
- Intended for qualitative use only
It consists of acetaldehyde, acetone, tert-butanol (TBA), ethanol (BAC), methanol and 2-propanol (isopropanol).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.