Order Entry
Germany
ContactUsLinkComponent
Aldehyde-ketone-DNPH TO-11A calibration mix (15 components)
Aldehyde-ketone-DNPH TO-11A calibration mix (15 components)
Catalog # REST31808
Supplier:  Restek
CAS Number:  
undefined
Aldehyde-ketone-DNPH TO-11A calibration mix (15 components)
Catalog # REST31808
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    15 µg/ml each in acetonitrile
  • Pk:
    1 Ampoul
  • Description:
    Aldehyde-ketone-DNPH TO-11A, calibration mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

This consists of acetaldehyde-DNPH; acetone-DNPH; acrolein-DNPH; benzaldehyde-DNPH; n-butyraldehyde-DNPH; crotonaldehyde-DNPH; 2,5-dimethylbenzaldehyde-DNPH; formaldehyde-DNPH; hexaldehyde-DNPH; isovaleraldehyde-DNPH; propionaldehyde-DNPH; m-tolualdehyde-DNPH; o-Tolualdehyde-DNPH; p-tolualdehyde-DNPH and aleraldehyde-DNPH.

Caution: *The reported concentrations reflect the amount of aldehyde or ketone in the mixture. The concentration of derivatized aldehyde or ketone is not reported.